Learn More
Medchemexpress LLC Heptadecanoic acid | 506-12-7 | 270.45 | 25 G
SDP
Catalog No. 5000449472
Encompass_Preferred
Heptadecanoic acid is an odd-chain saturated fatty acid (OCS-FA) with oral activity. It can inhibit cell proliferation and induce apoptosis, demonstrating antitumor activity. It is associated with a number of diseases, including coronary heart disease, pre-diabetes, type 2 diabetes, and multiple sclerosis.
  • Inhibits cell proliferation and migration
  • Promotes apoptosis
  • Associated with various disease research fields
Catalog No. 50-004-49472 Supplier Medchemexpress LLC Supplier No. 25GHYW004284
Heptadecanoic acid is an odd-chain saturated fatty acid (OCS-FA) with oral activity. It can inhibit cell proliferation and induce apoptosis, demonstrating antitumor activity. It is associated with a number of diseases, including coronary heart disease, pre-diabetes, type 2 diabetes, and multiple sclerosis.
  • Inhibits cell proliferation and migration
  • Promotes apoptosis
  • Associated with various disease research fields