Learn More
Medchemexpress LLC Heptadecanoic acid | 506-12-7 | 270.45 | 5 G
SDP
Catalog No. 5000445733
Encompass_Preferred
Heptadecanoic acid is an odd-chain saturated fatty acid (OCS-FA) with oral activity. It can inhibit cell proliferation and induce apoptosis, exhibiting antitumor activity. It is associated with various diseases, including coronary heart disease, pre-diabetes and type 2 diabetes, and multiple sclerosis.
  • Classified as an endogenous metabolite
  • Appears solid below 59°C and liquid above 61°C
  • White to off-white color
  • Initial source is microorganisms
Catalog No. 50-004-45733 Supplier Medchemexpress LLC Supplier No. HYW0042845G
Heptadecanoic acid is an odd-chain saturated fatty acid (OCS-FA) with oral activity. It can inhibit cell proliferation and induce apoptosis, exhibiting antitumor activity. It is associated with various diseases, including coronary heart disease, pre-diabetes and type 2 diabetes, and multiple sclerosis.
  • Classified as an endogenous metabolite
  • Appears solid below 59°C and liquid above 61°C
  • White to off-white color
  • Initial source is microorganisms