Learn More
Medchemexpress LLC Heptadecanoic acid-d3 | 202528-95-8 | 273.47 | 10 MG
SDP
Catalog No. 5000445699
Encompass_Preferred
Heptadecanoic acid-d3 is the deuterium labeled Heptadecanoic acid. Heptadecanoic acid is an odd chain saturated fatty acid (OCS-FA). It is associated with several diseases, including coronary heart disease, prediabetes, type 2 diabetes, and multiple sclerosis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-45699 Supplier Medchemexpress LLC Supplier No. HYW004284S10MG
Heptadecanoic acid-d3 is the deuterium labeled Heptadecanoic acid. Heptadecanoic acid is an odd chain saturated fatty acid (OCS-FA). It is associated with several diseases, including coronary heart disease, prediabetes, type 2 diabetes, and multiple sclerosis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS