Learn More
Medchemexpress LLC Heptadecanoic acid-d33 | 352431-41-5 | 303.65 | 10 MG
SDP
Catalog No. 5000445765
Encompass_Preferred
Heptadecanoic acid-d33 is a deuterium labeled odd chain saturated fatty acid. It can be used as a tracer and serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled odd chain saturated fatty acid
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for drug development quantitation
  • Affects pharmacokinetic and metabolic profiles of drugs
  • Solid appearance, white to off-white
Catalog No. 50-004-45765 Supplier Medchemexpress LLC Supplier No. HYW004284S110MG
Heptadecanoic acid-d33 is a deuterium labeled odd chain saturated fatty acid. It can be used as a tracer and serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled odd chain saturated fatty acid
  • Used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for drug development quantitation
  • Affects pharmacokinetic and metabolic profiles of drugs
  • Solid appearance, white to off-white