Learn More
Medchemexpress LLC Heptadecanoic acid-d33 | 352431-41-5 | 303.65 | 100 MG
SDP
Catalog No. 5000445764
Encompass_Preferred
Heptadecanoic acid-d33 is the deuterium labeled Heptadecanoic acid. Heptadecanoic acid is an odd chain saturated fatty acid (OCS-FA). Heptadecanoic acid is associated with several diseases, including the incidence of coronary heart disease, prediabetes and type 2 diabetes as well as multiple sclerosis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-45764 Supplier Medchemexpress LLC Supplier No. HYW004284S1100MG
Heptadecanoic acid-d33 is the deuterium labeled Heptadecanoic acid. Heptadecanoic acid is an odd chain saturated fatty acid (OCS-FA). Heptadecanoic acid is associated with several diseases, including the incidence of coronary heart disease, prediabetes and type 2 diabetes as well as multiple sclerosis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs.