Learn More
Medchemexpress LLC Heptadecanoic acid-d33 | 352431-41-5 | 303.65 | 5 MG
SDP
Catalog No. 5000447515
Encompass_Preferred
Heptadecanoic acid-d33 is the deuterium labeled Heptadecanoic acid. Heptadecanoic acid is an odd chain saturated fatty acid (OCS-FA) and is associated with various diseases, including coronary heart disease, prediabetes, type 2 diabetes, and multiple sclerosis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-47515 Supplier Medchemexpress LLC Supplier No. HYW004284S5MG
Heptadecanoic acid-d33 is the deuterium labeled Heptadecanoic acid. Heptadecanoic acid is an odd chain saturated fatty acid (OCS-FA) and is associated with various diseases, including coronary heart disease, prediabetes, type 2 diabetes, and multiple sclerosis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.