Learn More
Cayman Chemical HeptadecnoIc AcId 500mg
SDP
Catalog No. 502984189
Encompass_Preferred
An odd-chain saturated fatty acid; has been found in milk fat; has been used as an internal standard for the quantification of fatty acids in human plasma by LC- and GC-MS; has been used as a biomarker for dairy fat intake
Catalog No. 50-298-4189 Supplier Cayman Chemical Supplier No. 19722500 MG
An odd-chain saturated fatty acid; has been found in milk fat; has been used as an internal standard for the quantification of fatty acids in human plasma by LC- and GC-MS; has been used as a biomarker for dairy fat intake