Learn More
Cayman Chemical Heptadecanoic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 202528-95-8, 10 mg
SDP
Catalog No. 502938935
Encompass_Preferred
An internal standard for the quantification of heptadecanoicacid by GC- or LC-MS
Catalog No. 50-293-8935 Supplier Cayman Chemical Supplier No. 2787010 MG
An internal standard for the quantification of heptadecanoicacid by GC- or LC-MS