Learn More
Cayman Chemical HeptadecnoIc AcId-d3 25mg
SDP
Catalog No. 502937545
Encompass_Preferred
An internal standard for the quantification of heptadecanoicacid by GC- or LC-MS
Catalog No. 50-293-7545 Supplier Cayman Chemical Supplier No. 2787025 MG
An internal standard for the quantification of heptadecanoicacid by GC- or LC-MS