Learn More
Cayman Chemical Heptadecanoyl-Coenzyme A (lithium salt hydrate), Purity: ≥95%, 1 mg
SDP
Catalog No. 502933363
Encompass_Preferred
A long-chain saturated fatty acyl CoA; has been used as an internal standard for the quantification of hepatic and mitochondrial fatty acyl CoAs
Catalog No. 50-293-3363 Supplier Cayman Chemical Supplier No. 259991 MG
A long-chain saturated fatty acyl CoA; has been used as an internal standard for the quantification of hepatic and mitochondrial fatty acyl CoAs