Learn More
Cayman Chemical Heptadecnoyl EthnolamIde 100mg
SDP
Catalog No. 502895012
Encompass_Preferred
A F508del-CFTR corrector increases CFTR levels in CFBE41o- bronchial epithelial cells expressing F508del-CFTR fused to HRP (EC50 0.13 UM) and the transepithelial current in primary human bronchial epithelial cells isolated from a patient with cystic fibrosis with the F508del-CFTR mutation (EC50 0.028 UM)
Catalog No. 50-289-5012 Supplier Cayman Chemical Supplier No. 90342100 MG
A F508del-CFTR corrector increases CFTR levels in CFBE41o- bronchial epithelial cells expressing F508del-CFTR fused to HRP (EC50 0.13 UM) and the transepithelial current in primary human bronchial epithelial cells isolated from a patient with cystic fibrosis with the F508del-CFTR mutation (EC50 0.028 UM)