Learn More
Cayman Chemical Heptadecanoyl Ethanolamide, Purity: ≥98%, CAS: 53832-59-0, 50 mg
SDP
Catalog No. 502894869
Encompass_Preferred
A synthetic analog of PEA which incorporates an odd-numbered (17-carbon) fatty acid achain; potentiates the Ca2+ influx in response to AEA several fold in cells expressing human recombinant VR1
Catalog No. 50-289-4869 Supplier Cayman Chemical Supplier No. 9034250 MG
A synthetic analog of PEA which incorporates an odd-numbered (17-carbon) fatty acid achain; potentiates the Ca2+ influx in response to AEA several fold in cells expressing human recombinant VR1