Learn More
Cayman Chemical Heptadecnoyl-L-carnItIn-d3 1mg
SDP
Supplier:  Cayman Chemical 90032631 MG
Encompass_Preferred
An internal standard for the quantification of heptadecanoyl-L-carnitine by GC- or LC-MS
Catalog No. 50-289-4201