Learn More
Cayman Chemical Heptadecnoyl-L-carnItInch 10mg
SDP
Catalog No. 502917676
Encompass_Preferred
A long-chain acylcarnitine; levels are increased in dried blood spots from neonates with propionic or methylmalonic acidemias
Catalog No. 50-291-7676 Supplier Cayman Chemical Supplier No. 3793410 MG
A long-chain acylcarnitine; levels are increased in dried blood spots from neonates with propionic or methylmalonic acidemias