Learn More
Cayman Chemical HeptadecnoylLcarnItInd3 10mg
SDP
Catalog No. 502894420
Encompass_Preferred
An internal standard for the quantification of heptadecanoyl-L-carnitine by GC- or LC-MS
Catalog No. 50-289-4420 Supplier Cayman Chemical Supplier No. 900326310 MG
An internal standard for the quantification of heptadecanoyl-L-carnitine by GC- or LC-MS