Learn More
Selleck Chemical LLC Heptaethylene glycol S1697-100mg
SDP
Catalog No. 503110037
Encompass_Preferred
Heptaethylene glycol (HO-PEG7-OH) is a PEG-based PROTAC linker that is applicable to the synthesis of PROTACs
Catalog No. 50-311-0037 Supplier Selleck Chemical LLC Supplier No. S1697100MG
Heptaethylene glycol (HO-PEG7-OH) is a PEG-based PROTAC linker that is applicable to the synthesis of PROTACs