Learn More
Medchemexpress LLC Heptaethylene glycol | 5617-32-3 | ≥97.0% | C14H30O8 | 10 G
SDP
Catalog No. 5000352946
Encompass_Preferred
Heptaethylene glycol is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound is intended for research use only. It appears as a colorless to off-white solid below 17.8°C and a liquid above 18.8°C, with a high purity of ≥97.0%.
  • Utilized in the synthesis of PROTACs
  • Has a molecular weight of 326.38
  • Stable under recommended storage conditions
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-52946 Supplier Medchemexpress LLC Supplier No. HY14123110G
Heptaethylene glycol is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound is intended for research use only. It appears as a colorless to off-white solid below 17.8°C and a liquid above 18.8°C, with a high purity of ≥97.0%.
  • Utilized in the synthesis of PROTACs
  • Has a molecular weight of 326.38
  • Stable under recommended storage conditions
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month