Learn More
Medchemexpress LLC Heptaethylene glycol | 5617-32-3 | ≥97.0% | C14H30O8 | 25 G
SDP
Catalog No. 5000352508
Encompass_Preferred
Heptaethylene glycol is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs function by connecting two ligands: one targets an E3 ubiquitin ligase, and the other targets a protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
  • Leverages the ubiquitin-proteasome system
  • Suitable for research applications only
Catalog No. 50-003-52508 Supplier Medchemexpress LLC Supplier No. 25GHY141231
Heptaethylene glycol is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs function by connecting two ligands: one targets an E3 ubiquitin ligase, and the other targets a protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
  • Leverages the ubiquitin-proteasome system
  • Suitable for research applications only