Learn More
Medchemexpress LLC Heptaethylene glycol | 5617-32-3 | ≥97.0% | C14H30O8 | 5 G
SDP
Catalog No. 5000355304
Encompass_Preferred
Heptaethylene glycol is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Can be used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand is for an E3 ubiquitin ligase
  • The other ligand is for the target protein
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55304 Supplier Medchemexpress LLC Supplier No. HY1412315G
Heptaethylene glycol is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Can be used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand is for an E3 ubiquitin ligase
  • The other ligand is for the target protein
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins