Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptaethylene Glycol 96.0+%, TCI America™
SDP

Catalog No. H10451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10451G Supplier TCI America Supplier No. H10451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5617-32-3
Molecular Formula C14H30O8
Molecular Weight (g/mol) 326.386
MDL Number MFCD00002876
InChI Key XPJRQAIZZQMSCM-UHFFFAOYSA-N
PubChem CID 79718
ChEBI CHEBI:44748
IUPAC Name 2-[2-[2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCOCCOCCOCCO)O

Specifications

Color Yellow
Boiling Point 244°C
Quantity 1 g
Formula Weight 326.39
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptaethylene Glycol
EINECSNumber (2)-0441
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.