Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptafluoro-2-iodopropane, 97%, stab. with copper powder

Catalog No. AAL1652306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1652306 5 g
AAL1652314 25 g
AAL1652322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1652306 Supplier Thermo Scientific Chemicals Supplier No. L1652306
Add to Cart

CAS: 677-69-0 | C3F7I | 295.926 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 677-69-0
Molecular Formula C3F7I
Molecular Weight (g/mol) 295.926
MDL Number MFCD00042089
InChI Key BBZVTTKMXRPMHZ-UHFFFAOYSA-N
Synonym heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide
PubChem CID 33978
IUPAC Name 1,1,1,2,3,3,3-heptafluoro-2-iodopropane
SMILES C(C(F)(F)F)(C(F)(F)F)(F)I

Specifications

Melting Point -56°C
Density 2.1
Boiling Point 38°C
Refractive Index 1.33
Linear Formula (CF3)2CFI
Quantity 5 g
Beilstein 1841228
Sensitivity Light sensitive
Formula Weight 295.93
Percent Purity 97%
Chemical Name or Material Heptafluoro-2-iodopropane, stabilized with copper powder
Show More Show Less
Hazard Category H315-H319-H332-H335
Hazard Statement GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-643-3
RTECSNumber TZ3925000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.