Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptafluorobutyramide 97.0+%, TCI America™
SDP

Catalog No. H092625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H092625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H092625G Supplier TCI America Supplier No. H092625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 662-50-0
Molecular Formula C4H2F7NO
Molecular Weight (g/mol) 213.055
MDL Number MFCD00039773
InChI Key FOBJABJCODOMEO-UHFFFAOYSA-N
Synonym heptafluorobutyramide, perfluorobutyramide, butanamide, 2,2,3,3,4,4,4-heptafluoro, heptafluorobutanamide, heptafluorobutyroamide, heptafluoro-n-butyramide, butyramide, 2,2,3,3,4,4,4-heptafluoro, 2,2,3,3,4,4,4-heptafluorobutyramide, 2,2,3,3,4,4,4-heptafluoro-butyramide, butyramide, alpha,alpha,beta,beta,gamma,gamma,gamma-heptafluoro
PubChem CID 12626
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanamide
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)N

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 25 g
Formula Weight 213.05
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Heptafluorobutyramide
RTECSNumber ES4725000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.