Learn More
Cayman Chemical Heptafluorobutyric Acid, Purity: ≥80%, CAS: 375-22-4, 10 g
SDP
Catalog No. 502915403
Encompass_Preferred
A perfluoroalkyl substance; chronic topical administration on the ear increases liver and kidney weight in mice at 25 µl of a 3.75-15% v/v on the ear; induces multifocal hepatocyte necrosis and centrilobular hepatocyte hypertrophy at 7.5% v/v; increases serum cholesterol and ALP levels at 15% v/v; decreases serum urea nitrogen levels; has been found in marine life and as a contaminant in surface water
Catalog No. 50-291-5403 Supplier Cayman Chemical Supplier No. 3723410 G
A perfluoroalkyl substance; chronic topical administration on the ear increases liver and kidney weight in mice at 25 µl of a 3.75-15% v/v on the ear; induces multifocal hepatocyte necrosis and centrilobular hepatocyte hypertrophy at 7.5% v/v; increases serum cholesterol and ALP levels at 15% v/v; decreases serum urea nitrogen levels; has been found in marine life and as a contaminant in surface water