Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Heptafluorobutyric acid

Catalog No. 164194100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-419-4100G 100 g
16-419-425G 25 g
16-419-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-419-4100G Supplier Sigma Aldrich Supplier No. 164194100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Perfluorobutyric acid is an effective additive for zinc electrodeposition.perfluorobutyric acid was used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins
  • It was used as ion pairing reagent in preparative and analytical reserved phase HPLC of underivatised peptides

Specifications

CAS 375-22-4
Density 1.645 g/mL (at 25°C (literature))
Boiling Point 120°C (755 mmHg)
Molecular Formula C4HF7O2
Refractive Index n20/D 1.3 (literature)
Linear Formula CF3CF2CF2COOH
MDL Number MFCD00004171
Quantity 100 g
Synonym Edman Reagent No. 3; HFBA; Perfluorobutyric acid
Molecular Weight (g/mol) 214.04
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.