Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptafluorobutyric Acid, 99.6%, MP Biomedicals™

Catalog No. ICN15123725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15123725 25 mL
ICN15123710 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15123725 Supplier MP Biomedicals, Inc Supplier No. 0215123725
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heptafluorobutyric Acid is used in determination of amino acid sequences in proteins.

  • Heptafluorobutyric Acid is used as a mobile phase modifier which significantly improved selectivity in the HPLC analysis of histone proteins.
  • It can be used at a concentration of 0.1% in the mobile phase of an HPLC/LC-MS protocol for the detection of marine bacterioplankton siderophores.
  • It is used in the chiral LC analysis of (+)- and (-)-epibatidine.
  • The analysis of selenium containing compounds using HFBA in the MS matrix has been described.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4HF7O2
Molecular Weight (g/mol) 214.039
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
Synonym heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
PubChem CID 9777
ChEBI CHEBI:39426
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O

Specifications

Color Colorless
Density 1.645 g/mL
Assay Percent Range 99.6%
Quantity 25 mL
Formula Weight 214.04
Physical Form Liquid
Chemical Name or Material Heptafluorobutyric acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.