Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptafluorobutyric anhydride, 98%

Catalog No. AAA1074422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1074422 100 g
AAA1074406 5 g
AAA1074414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1074422 Supplier Thermo Scientific Chemicals Supplier No. A1074422
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 336-59-4 | C8F14O3 | 410.06 g/mol

Heptafluorobutyric anhydride is used as a derivatization reagent for gas chromatography - mass mass spectrometry analysis of the stereoisomer content of synthetic peptides. It is a GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptafluorobutyric anhydride is used as a derivatization reagent for gas chromatography - mass mass spectrometry analysis of the stereoisomer content of synthetic peptides. It is a GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.

Solubility
Miscible with halogenated solvents. Immiscible with polar solvents.

Notes
Incompatible with moist air, water, strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336-59-4
Molecular Formula C8F14O3
Molecular Weight (g/mol) 410.06
MDL Number MFCD00000432
InChI Key UFFSXJKVKBQEHC-UHFFFAOYSA-N
Synonym heptafluorobutyric anhydride, perfluorobutyric anhydride, hfba, heptafluorobutanoic anhydride, 2,2,3,3,4,4,4-heptafluorobutanoic anhydride, butanoic acid, heptafluoro-, anhydride, heptafluoro-n-butyric anhydride, heptafluorobutyric acid anhydride, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, 1,1'-anhydride, heptafluorobutyric anhydride, for gc derivatization
PubChem CID 67643
ChEBI CHEBI:39424
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point -43°C
Density 1.662
Boiling Point 108°C to 110°C
Odor Stench
Refractive Index 1.287
Linear Formula (CF3CF2CF2CO)2O
Quantity 100 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Miscible with halogenated solvents. Immiscible with polar solvents.
Formula Weight 410.06
Percent Purity 98%
Chemical Name or Material Heptafluorobutyric anhydride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-410-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.