Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Heptakis(2,6-Di-O-Methyl)-Beta-Cyclodextrin

Catalog No. H05131G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05131G 5 g
50-308-1498 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05131G Supplier Sigma Aldrich Supplier No. H05131G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Heptakis(2,6-di-O-methyl)-+a-cyclodextrin is a hydrophilic derivative of cyclodextrin that possesses the ability to effectively dissolve lipophilic drugs like steroid hormones, vitamins A, E, and K, etc. It can be utilized to increase the dissolution and bioavailability of weakly water-soluble pharmaceutical compounds.Reactant involved in:Physicochemical studies of inclusion complexesProbing diffusion and single molecule interactions with reconstituted membrane proteinsInteractions with micelles causing micellar ruptureStudying cellular transport, used as a cell penetration enhancerPhysicochemical and biopharmaceutical improvement of drugs particularly with solubilization and stabilityInteractions with cholesterol

Specifications

CAS 51166-71-3
Molecular Formula C56H98O35
Linear Formula C56H98O35
MDL Number MFCD00011616
Quantity 5 g
Synonym 2,6-Di-O-methyl-beta-cyclodextrin; Dimethyl beta-cyclodextrin
Molecular Weight (g/mol) 1331.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.