Learn More
Medchemexpress LLC HY-D0921 5mg Medchemexpress, Heptamethine cyanine dye-1 CAS:162411-29-2 Purity:>98%
SDP
Catalog No. 501933209
Encompass_Preferred
Medchemexpress, HY-D0921 5mg Heptamethine cyanine dye-1 CAS:162411-29-2 Heptamethine cyanine dye-1 is a near-infrared cyanine dye for fluorescence imaging in biological systems. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3209 Supplier Medchemexpress LLC Supplier No. HYD09215MG
Medchemexpress, HY-D0921 5mg Heptamethine cyanine dye-1 CAS:162411-29-2 Heptamethine cyanine dye-1 is a near-infrared cyanine dye for fluorescence imaging in biological systems. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.