Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptamethyldisilazane, 99%, Thermo Scientific™

Catalog No. AC215920100
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC215920100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC215920100 Supplier Thermo Scientific Chemicals Supplier No. 215920100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Silylation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 920-68-3
Density 0.7900g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C7H21NSi2
Refractive Index 1.4200 to 1.4220 (20°C, 589nm)
Linear Formula (CH3)3SiN(CH3)Si(CH3)3
Quantity 10g
MDL Number MFCD00008262
Synonym heptamethyldisilazane, n,1,1,1-tetramethyl-n-trimethylsilyl silanamine, silanamine, n,1,1,1-tetramethyl-n-trimethylsilyl, heptamethyl disilazane, n,n-bis trimethylsilyl methylamine, methylbis trimethylsilyl amine, methylamine, 2tms derivative, n,n-bis trimethylsilyl methanamine, n,1,1,1-tetramethyl-n-trimethylsilyl silylamine, acmc-1c1go
InChI Key ZSMNRKGGHXLZEC-UHFFFAOYSA-N
SMILES CN([Si](C)(C)C)[Si](C)(C)C
IUPAC Name N,N-bis(trimethylsilyl)methanamine
Molecular Weight (g/mol) 175.422
PubChem CID 70199
Formula Weight 175.43
Percent Purity ≥98.5%
Chemical Name or Material Heptamethyldisilazane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-061-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.