Learn More
Medchemexpress LLC HY-16918A 5mg Medchemexpress, Heptamidine (dimethanesulfonate) CAS:161374-55-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16918A5MG
Encompass_Preferred
Medchemexpress, HY-16918A 5mg Heptamidine (dimethanesulfonate) CAS:161374-55-6 Heptamidine dimethanesulfonate (SBi4211 dimethanesulfonate) is a potent Pentamidine-related inhibitor of the calcium-binding protein S100B (Kd=6.9 μM), selectively kills melanoma cells with S100B over those without S100B. Heptamidine is a useful tool for the investigation of Myotonic dystrophy (DM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1916