Learn More
Medchemexpress LLC Heptamidine dimethanesulfonate | 161374-55-6 | 99.8% | 560.68 | 100 MG
SDP
Catalog No. 5000390540
Encompass_Preferred
Heptamidine dimethanesulfonate, also known as SBi4211 dimethanesulfonate, is a potent Pentamidine-related inhibitor of the calcium-binding protein S100B with a Kd of 6.9 μM. It selectively eliminates melanoma cells that express S100B, while sparing those without it. This compound also serves as a valuable tool for myotonic dystrophy (DM) research, demonstrating the ability to rescue mis-splicing of minigene reporters in a HeLa cell DM1 model with an EC50 value of 15 μM and reversing splicing defects and myotonia in a DM mouse model. The product has a purity of 99.82%, a CAS number of 161374-55-6, and a molecular weight of 560.68.
  • Potent S100B inhibitor
  • Selective melanoma cell elimination (S100B-positive)
  • Useful tool for myotonic dystrophy (DM) investigation
  • Reverses splicing defects and myotonia in DM models
  • High purity: 99.82%
  • Molecular weight: 560.68
  • Off-white to light yellow solid
Catalog No. 50-003-90540 Supplier Medchemexpress LLC Supplier No. HY16918A100MG
Heptamidine dimethanesulfonate, also known as SBi4211 dimethanesulfonate, is a potent Pentamidine-related inhibitor of the calcium-binding protein S100B with a Kd of 6.9 μM. It selectively eliminates melanoma cells that express S100B, while sparing those without it. This compound also serves as a valuable tool for myotonic dystrophy (DM) research, demonstrating the ability to rescue mis-splicing of minigene reporters in a HeLa cell DM1 model with an EC50 value of 15 μM and reversing splicing defects and myotonia in a DM mouse model. The product has a purity of 99.82%, a CAS number of 161374-55-6, and a molecular weight of 560.68.
  • Potent S100B inhibitor
  • Selective melanoma cell elimination (S100B-positive)
  • Useful tool for myotonic dystrophy (DM) investigation
  • Reverses splicing defects and myotonia in DM models
  • High purity: 99.82%
  • Molecular weight: 560.68
  • Off-white to light yellow solid