Learn More
Sigma Aldrich Fine Chemicals Biosciences Heptane anhydrous 99
Catalog No. 502898090
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent and greener alternative to hexane and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.Heptane is a linear hydrocarbon that is considered as a greener alternative to the more toxic hexane.
Catalog No. 50-289-8090 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2466544X2L
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent and greener alternative to hexane and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.Heptane is a linear hydrocarbon that is considered as a greener alternative to the more toxic hexane.