Learn More
Sigma Aldrich Fine Chemicals Biosciences Heptane, 142-82-5, MFCD00009544, 2 L
Catalog No. NC1214206
Encompass
Linear Formula: CH3(CH2)5CH3, Molecular Weight: 100.20, anhydrous, 99%. Heptane is a linear hydrocarbon that is considered as a greener alternative to the more toxic hexane.
Catalog No. NC1214206 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2466542L
Linear Formula: CH3(CH2)5CH3, Molecular Weight: 100.20, anhydrous, 99%. Heptane is a linear hydrocarbon that is considered as a greener alternative to the more toxic hexane.