Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (>99.0% n-Heptane), Solstice

Catalog No. 6001033 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-010-33 6 x 1 L Glass Bottle
60-010-32 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-010-33 Supplier Solstice Supplier No. 2071L
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Absorbance 0.1 AU max. at 225 nm (UV), 0.01 AU max. at 250 nm (UV), 0.005 AU max. at 300 nm (UV)
CAS Max % 100.0000%
Density 0.684 g/cm3
Quantity 6 x 1 L
Flash Point −4°C
Packaging Glass Bottle
UN Number UN1206
Beilstein 1730763
Boiling Point 98.4°C
Formula Weight 100.21
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Heptane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (>99.0% n-Heptane)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.