Learn More
Sigma Aldrich Fine Chemicals Biosciences Heptane HPLC Plus for HPLC1L
Catalog No. 502903604
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent and greener alternative to hexane for HPLC and GC and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.Heptane (n-Heptane) is a primary reference fuel (PRf) for the rating of octane numbers of fuels in internal combustion engines. Its cetane number has been reported to be 56. Mechanism and kinetic studies of its oxidation in flow reactors shock tubes and rapid compression machines have been proposed. Its thermal decomposition has been studied under different conditions of temperature and pressure. Its oxidative dehydrogenation in the presence of magnesium oxide supported vanadium catalyst has been investigated. Molybdenum phosphide supported on H zeolite (MoP/H) catalyzed hydroisomerization of n-heptane has been reported to be enhanced by doping with secondary
Catalog No. 50-290-3604 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6505366X1L
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent and greener alternative to hexane for HPLC and GC and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.Heptane (n-Heptane) is a primary reference fuel (PRf) for the rating of octane numbers of fuels in internal combustion engines. Its cetane number has been reported to be 56. Mechanism and kinetic studies of its oxidation in flow reactors shock tubes and rapid compression machines have been proposed. Its thermal decomposition has been studied under different conditions of temperature and pressure. Its oxidative dehydrogenation in the presence of magnesium oxide supported vanadium catalyst has been investigated. Molybdenum phosphide supported on H zeolite (MoP/H) catalyzed hydroisomerization of n-heptane has been reported to be enhanced by doping with secondary