Learn More
Sigma Aldrich Fine Chemicals Biosciences Heptane suitable for HPLC
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 348734X4L
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent and greener alternative to hexane for chromatographic purification and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.Tools and techniques for solvent selection green solvent selection guides.Heptane (n-Heptane) is a linear hydrocarbon. Its cetane number has been reported to be 56.
Catalog No. 50-290-3478