Learn More
Sigma Aldrich Fine Chemicals Biosciences Heptane suitable for HPLC
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 5925794X4L
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent and greener alternative to hexane for chromatographic purification and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.Tools and techniques for solvent selection green solvent selection guides.Heptane (n-Heptane) is a primary reference fuel (PRF) for the rating of octane numbers of fuels in internal combustion engines. Its cetane number has been reported to be 56. Mechanism and kinetic studies of its oxidation in flow reactors shock tubes and rapid compression machines have been proposed. Its thermal decomposition has been studied under different conditions of temperature and pressure. Its oxidative dehydrogenation in the presence of magnesium oxide supported vanadium catalyst has been investigated. Molybdenum phosphide supported on H zeolite (MoP/H) catalyze
Catalog No. 50-290-7842