Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptanenitrile, 98%, Thermo Scientific™

Catalog No. AC309340250
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC309340250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC309340250 Supplier Thermo Scientific Chemicals Supplier No. 309340250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-08-3
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.19
MDL Number MFCD00039503
InChI Key SDAXRHHPNYTELL-UHFFFAOYSA-N
Synonym heptanonitrile, hexyl cyanide, enanthonitrile, 1-cyanohexane, 1-heptanonitrile, heptane-1-nitrile, n-heptanenitrile, unii-ua4z3qt98a, ua4z3qt98a, heptane nitrile
PubChem CID 12372
IUPAC Name heptanenitrile
SMILES CCCCCCC#N

Specifications

Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 mL
Formula Weight 111.19
Chemical Name or Material Heptanenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.