Learn More
Medchemexpress LLC Heptanoate (sodium) | 10051-45-3 | 99.9% | 152.17 | 10 G
SDP
Catalog No. 5000403956
Encompass_Preferred
Heptanoate sodium is an organic sodium salt compound widely used in various industries and laboratories. It can be utilized as a surfactant, emulsifier, lubricant, and preservative. It also plays a role in some electronic devices and in the preparation of chemicals and raw materials like paints, plastics, and fragrances. While it has no direct medical application, it is important in chemical research and industrial production. As a biochemical reagent, it can be used as a biological material or organic compound for life science related research.
  • Used as a surfactant
  • Used as an emulsifier
  • Used as a lubricant
  • Used as a preservative
  • Important in chemical research
  • Important in industrial production
  • Biochemical reagent for life science related research
Catalog No. 50-004-03956 Supplier Medchemexpress LLC Supplier No. HY42935A10G
Heptanoate sodium is an organic sodium salt compound widely used in various industries and laboratories. It can be utilized as a surfactant, emulsifier, lubricant, and preservative. It also plays a role in some electronic devices and in the preparation of chemicals and raw materials like paints, plastics, and fragrances. While it has no direct medical application, it is important in chemical research and industrial production. As a biochemical reagent, it can be used as a biological material or organic compound for life science related research.
  • Used as a surfactant
  • Used as an emulsifier
  • Used as a lubricant
  • Used as a preservative
  • Important in chemical research
  • Important in industrial production
  • Biochemical reagent for life science related research