Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heptanoic acid, 2-propen-1-yl ester | 142-19-8 | MFCD00038341 | 99.8% | 170.25 g/mol | C10H18O2 | 25 G
SDP

Catalog No. 5000303098
Encompass_Preferred

Allyl heptanoate (heptanoic acid, 2-propen-1-yl ester) is a laboratory reagent ester supplied as a liquid for biochemical and research use. It offers consistent purity and well-documented solubility and storage characteristics to support reproducible experiments.

  • High purity (99.8%) providing reagent-grade quality.
  • Liquid, colorless to light yellow, density 0.885 g/cm3.
  • Molecular weight 170.25 g/mol; formula C10H18O2.
  • Highly soluble in DMSO (100 mg/mL); in vivo formulation protocols reported.
  • Available in small lab sizes suitable for bench work (5 g, 25 g, 50 g).
  • Stable when stored under recommended conditions (e.g., -20°C to 4°C depending on form).

Catalog No. 50-003-03098 Supplier Medchemexpress LLC Supplier No. 25GHYW016976
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allyl heptanoate (heptanoic acid, 2-propen-1-yl ester) is a laboratory reagent ester supplied as a liquid for biochemical and research use. It offers consistent purity and well-documented solubility and storage characteristics to support reproducible experiments.

  • High purity (99.8%) providing reagent-grade quality.
  • Liquid, colorless to light yellow, density 0.885 g/cm3.
  • Molecular weight 170.25 g/mol; formula C10H18O2.
  • Highly soluble in DMSO (100 mg/mL); in vivo formulation protocols reported.
  • Available in small lab sizes suitable for bench work (5 g, 25 g, 50 g).
  • Stable when stored under recommended conditions (e.g., -20°C to 4°C depending on form).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.