Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heptanoic acid, 2-propen-1-yl ester | 142-19-8 | MFCD00038341 | 99.8% | 170.25 g/mol | C10H18O2 | 5 G
SDP

Catalog No. 5000301263
Encompass_Preferred

Allyl heptanoate (Prop-2-en-1-yl heptanoate) is a medium-chain fatty acid ester supplied as a high-purity biochemical reagent. It is a colorless to light yellow liquid with a fruity odor, commonly used as an organic intermediate and flavoring agent in research. Molecular formula C10H18O2; molecular weight 170.25 g/mol.

  • High purity: 99.8%.
  • Appearance: colorless to light yellow liquid; density 0.885 g/cm3.
  • Molecular formula C10H18O2; molecular weight 170.25 g/mol.
  • Common applications: organic synthesis, flavoring research, biochemical assays.
  • Available in small laboratory quantities (5 g) with SDS and COA documentation.

Catalog No. 50-003-01263 Supplier Medchemexpress LLC Supplier No. HYW0169765G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allyl heptanoate (Prop-2-en-1-yl heptanoate) is a medium-chain fatty acid ester supplied as a high-purity biochemical reagent. It is a colorless to light yellow liquid with a fruity odor, commonly used as an organic intermediate and flavoring agent in research. Molecular formula C10H18O2; molecular weight 170.25 g/mol.

  • High purity: 99.8%.
  • Appearance: colorless to light yellow liquid; density 0.885 g/cm3.
  • Molecular formula C10H18O2; molecular weight 170.25 g/mol.
  • Common applications: organic synthesis, flavoring research, biochemical assays.
  • Available in small laboratory quantities (5 g) with SDS and COA documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.