Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heptanoic acid, 7-[[(1,1-dimethylethoxy)carbonyl]amino]- | 60142-89-4 | ≥98.0% | 245.32 | 10 G
SDP

Catalog No. 5000453796
Encompass_Preferred

N-Boc-7-aminoheptanoic acid is a chemical compound with molecular formula C12H23NO4 and molecular weight 245.32. It appears as a white to off-white solid with a purity of ≥98.0%. This compound serves as an important building block in peptide synthesis and PROTAC technology.

  • White to off-white solid appearance.
  • Purity of ≥98.0%.
  • Molecular weight of 245.32.
  • Molecular formula C12H23NO4.
  • Used in peptide synthesis and PROTAC technology.

Catalog No. 50-004-53796 Supplier Medchemexpress LLC Supplier No. HYW01200110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Boc-7-aminoheptanoic acid is a chemical compound with molecular formula C12H23NO4 and molecular weight 245.32. It appears as a white to off-white solid with a purity of ≥98.0%. This compound serves as an important building block in peptide synthesis and PROTAC technology.

  • White to off-white solid appearance.
  • Purity of ≥98.0%.
  • Molecular weight of 245.32.
  • Molecular formula C12H23NO4.
  • Used in peptide synthesis and PROTAC technology.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.