Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heptanoic acid, 7-[[(1,1-dimethylethoxy)carbonyl]amino]- | 60142-89-4 | ≥98.0% | 245.32 | 5 G
SDP

Catalog No. 5000453793
Encompass_Preferred

N-Boc-7-aminoheptanoic acid is a chemical compound with a molecular weight of 245.32 and a purity of ≥98.0%. It appears as a white to off-white solid and is suitable for research use only.

  • Molecular formula: C12H23NO4
  • Molecular weight: 245.32
  • Appearance: White to off-white solid
  • Purity: ≥98.0%
  • Storage at room temperature for 3 years
  • Storage in solvent at -80°C for 2 years, -20°C for 1 year
  • For research use only

Catalog No. 50-004-53793 Supplier Medchemexpress LLC Supplier No. HYW0120015G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Boc-7-aminoheptanoic acid is a chemical compound with a molecular weight of 245.32 and a purity of ≥98.0%. It appears as a white to off-white solid and is suitable for research use only.

  • Molecular formula: C12H23NO4
  • Molecular weight: 245.32
  • Appearance: White to off-white solid
  • Purity: ≥98.0%
  • Storage at room temperature for 3 years
  • Storage in solvent at -80°C for 2 years, -20°C for 1 year
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.