Learn More
Medchemexpress LLC Heptanoic acid, 7-[[(1,1-dimethylethoxy)carbonyl]amino]- | 60142-89-4 | HY-W012001 | >=98.0% | 245.32 | 100 G
SDP
Catalog No. 5000454000
Encompass_Preferred
N-Boc-7-aminoheptanoic acid is a PROTAC linker used in the synthesis of PROTACs and other conjugation applications. It features an alkane chain with terminal carboxylic acid and Boc-protected amino groups. The terminal carboxylic acid can react with primary amine groups in the presence of activators like EDC or HATU to form a stable amide bond, and the Boc group can be deprotected under mild acidic conditions to yield the free amine.
  • Utilized as a PROTAC linker
  • Suitable for PROTAC synthesis
  • Suitable for other conjugation applications
  • Features an alkane chain with terminal carboxylic acid and Boc-protected amino groups
  • Carboxylic acid reacts with primary amines for stable amide bond formation
  • Boc group deprotects under mild acidic conditions to yield free amine
Catalog No. 50-004-54000 Supplier Medchemexpress LLC Supplier No. HYW012001100G
N-Boc-7-aminoheptanoic acid is a PROTAC linker used in the synthesis of PROTACs and other conjugation applications. It features an alkane chain with terminal carboxylic acid and Boc-protected amino groups. The terminal carboxylic acid can react with primary amine groups in the presence of activators like EDC or HATU to form a stable amide bond, and the Boc group can be deprotected under mild acidic conditions to yield the free amine.
  • Utilized as a PROTAC linker
  • Suitable for PROTAC synthesis
  • Suitable for other conjugation applications
  • Features an alkane chain with terminal carboxylic acid and Boc-protected amino groups
  • Carboxylic acid reacts with primary amines for stable amide bond formation
  • Boc group deprotects under mild acidic conditions to yield free amine