Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptanoic Acid, MP Biomedicals

Catalog No. ICN210732 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210732 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210732 Supplier MP Biomedicals, Inc Supplier No. 0521073280

Chemical Identifiers

CAS 111-14-8
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
InChI Key MNWFXJYAOYHMED-UHFFFAOYSA-N
Synonym enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
PubChem CID 8094
ChEBI CHEBI:45571
IUPAC Name heptanoic acid
SMILES CCCCCCC(=O)O

Specifications

Quantity 100 g
Formula Weight 130.19
Chemical Name or Material Heptanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.