Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Heptanoyl chloride

Catalog No. 14724925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-724-925G 25 g
14-724-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-724-925G Supplier Sigma Aldrich Supplier No. 14724925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Heptanoyl chloride reacts with 2-ethylthiophene in the presence of stannic chloride to give 2-ethyl-5-heptanoylthiophene.heptanoyl chloride was used in synthesis of 5α-dihydrotestosterone heptanoate and α-ketoaldehyde
  • It was used in the preparation of ester-linked bilayer films

Specifications

CAS 2528-61-2
Density 0.96 g/mL (at 25°C (literature))
Boiling Point 173°C (lit.)
Molecular Formula C7H13ClO
Refractive Index n20/D 1.43 (literature)
Linear Formula CH3(CH2)5 COCl
MDL Number MFCD00000763
Quantity 25 g
Synonym Enanthic chloride; Oenanthic chloride
Molecular Weight (g/mol) 148.63
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.