Learn More
Cayman Chemical Heptanoyl thioPC
SDP
Supplier:  Cayman Chemical 100068095 MG
Encompass_Preferred
A thioester phospholipase substrate; exhibits 10-13 fold less activity for the porcine pancreatic and bee venom sPLA2 enzymes compared to diheptanoyl thio-PC
Catalog No. 50-288-9935