Learn More
Cayman Chemical Heptanoyl thioPC
SDP
Catalog No. 502890962
Encompass_Preferred
A thioester phospholipase substrate; exhibits 10-13 fold less activity for the porcine pancreatic and bee venom sPLA2 enzymes compared to diheptanoyl thio-PC
Catalog No. 50-289-0962 Supplier Cayman Chemical Supplier No. 1000680950 MG
A thioester phospholipase substrate; exhibits 10-13 fold less activity for the porcine pancreatic and bee venom sPLA2 enzymes compared to diheptanoyl thio-PC