Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptatriacontane 95.0+%, TCI America™
SDP

Catalog No. H09205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09205G 5 g
H09201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09205G Supplier TCI America Supplier No. H09205G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7194-84-5
Molecular Formula C37H76
Molecular Weight (g/mol) 521.015
MDL Number MFCD00042945
InChI Key PXEZIKSRSYGOED-UHFFFAOYSA-N
PubChem CID 23598
IUPAC Name heptatriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 77°C
Color White
Quantity 5 g
Formula Weight 521.02
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Heptatriacontane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.