Learn More
AdipoGen Heptenophos
SDP
Supplier:  AdipoGen CDXH0095M100
Encompass_Preferred
Chemical. CAS 23560-59-0. Formula C9H12ClO4P. MW 250. Synthetic. Synthetic insecticides derived from phosphoric acid, and capable of disrupting neurotransmission in insects and vertebrates. Ectoparasitocidal drug that works as an acetylcholine esterase inhibitor in arthropods. Compound can be used as analytical reference material.
Catalog No. 50-256-0746